missing translation for 'onlineSavingsMsg'
Learn More

HABP4 Antibody, Novus Biologicals™

Product Code. 18397278 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397278 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397278 Supplier Novus Biologicals Supplier No. H00022927D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HABP4 Polyclonal antibody specifically detects HABP4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HABP4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH18788.1
Gene Alias chromodomain helicase DNA binding protein 3 interacting protein, hyaluronan binding protein 4, IHABP-4, IHABP4MGC825, intracellular antigen detected by monoclonal Ki-1, intracellular hyaluronan-binding protein 4, Ki-1/57, Ki-1/57 intracellular antigen, SERBP1L
Host Species Rabbit
Immunogen HABP4 (AAH18788.1, 1 a.a. - 308 a.a.) full-length human protein. MKGALGSPVAAAGAAMQESFGCVVANRFHQLLDDESDPFDILREAERRRQQQLQRKRRDEAAAAAGAGPRGGRSPAGASGHRAGAGGRRESQKERKSLPAPVAQRPDSPGGGLQAPGQKRTPRRGEQQGWNDSRGPEGMLERAERRSYREYRPYETERQADFTAEKFPDEKVPELEVEEETQVQEMTLDEWKNLQEQTRPKPEFNIRKPESTVPSKAVVIHKSKYRDDMVKDDYEDDSHVFRKPANDITSQLEINFGNLPRPGRGARGGTRGGRGRIRRAENYGPRAEVVMQDVAPNPDDPEDFPALS
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22927
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.