missing translation for 'onlineSavingsMsg'
Learn More

HABP2 Antibody (OTI8D6) [Alexa Fluor™ 750], Novus Biologicals™

Product Code. 30689174 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689174 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689174 Supplier Novus Biologicals Supplier No. NBP344731AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HABP2 Monoclonal antibody specifically detects HABP2 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HABP2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI8D6
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.92, Factor seven-activating protease, Factor VII-activating protease, FSAPfactor VII activating protein, HABP, Hepatocyte growth factor activator-like protein, HGFALhyaluronic acid binding protein 2, hyaluronan binding protein 2, hyaluronan-binding protein 2, PHBPplasma hyaluronan binding protein, Plasma hyaluronan-binding protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 33-330 of human HABP2 (NP_004123) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3026
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.