missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HABP2 Antibody (OTI2B5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00
Specifications
| Antigen | HABP2 |
|---|---|
| Clone | OTI2B5 |
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306242
|
Novus Biologicals
NBP3-44942-100ul |
100 μL |
N/A
|
N/A | |||||
|
30306664
|
Novus Biologicals
NBP3-44942-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HABP2 Monoclonal antibody specifically detects HABP2 in Human samples. It is validated for Western BlotSpecifications
| HABP2 | |
| Western Blot 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 3026 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI2B5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.92, Factor seven-activating protease, Factor VII-activating protease, FSAPfactor VII activating protein, HABP, Hepatocyte growth factor activator-like protein, HGFALhyaluronic acid binding protein 2, hyaluronan binding protein 2, hyaluronan-binding protein 2, PHBPplasma hyaluronan binding protein, Plasma hyaluronan-binding protein | |
| Human recombinant protein fragment corresponding to amino acids 33-330 of human HABP2(NP_004123) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title