missing translation for 'onlineSavingsMsg'
Learn More

HABP1/C1QBP/GC1q R Rabbit anti-Mouse, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody

£286.00 - £429.00

Specifications

Antigen HABP1/C1QBP/GC1q R
Clone 1
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18294847
View Documents
Novus Biologicals
NBP2-90475-50UL
50 μg
£286.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18201987
View Documents
Novus Biologicals
NBP2-90475-100UL
100 μg
£429.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HABP1/C1QBP/GC1q R Monoclonal specifically detects HABP1/C1QBP/GC1q R in Mouse samples. It is validated for ELISA.
Specifications

Specifications

HABP1/C1QBP/GC1q R
ELISA 1:5000-1:10000
Monoclonal
Purified
RUO
Mouse
C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein
Recombinant Mouse HABP1/C1QBP/GC1q R protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
1
ELISA
Unconjugated
Rabbit
Stem Cell Markers
0.2 um filtered solution in PBS
708
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.