missing translation for 'onlineSavingsMsg'
Learn More

HABP1/C1QBP/GC1q R Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30734885 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734885 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734885 Supplier Novus Biologicals Supplier No. AF5359MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

HABP1/C1QBP/GC1q R Polyclonal antibody specifically detects HABP1/C1QBP/GC1q R in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HABP1/C1QBP/GC1q R
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein
Host Species Goat
Immunogen E. coli-derived recombinant mouse HABP1/C1QBP , Leu72-Gln279, Accession # NP_031599
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 708
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.