missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAAO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581604
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat HAAO/3-HAO.
HAAO (3-Hydroxyanthranilate 3, 4-dioxygenase) is a monomeric cytosolic protein of the family of intramolecular dioxygenases containing non-heme ferrous iron. It is widely distributed in peripheral organs, such as liver and kidney, and is present in low amounts in the central nervous system. This enzyme participates in tryptophan metabolism. It employs one cofactor, iron. HAAO catalyzes the synthesis of quinolinic acid (QUIN) from 3-hydroxyanthranilic acid. QUIN is an excitotoxin whose toxicity is mediated by its ability to activate glutamate N-methyl-D-aspartate receptors. Increased cerebral levels of QUIN may participate in the pathogenesis of neurological and inflammatory disorders. HAAO has been suggested to play a role in disorders associated with altered tissue levels of QUIN. Furthermore, recent study shows that HAAO are excellent candidate biomarkers for detecting ovarian cancer.
Specifications
| HAAO | |
| Polyclonal | |
| Unconjugated | |
| HAAO | |
| 0610007K21Rik; 0610012J07Rik; 3HAO; 3-HAO; 3-HAO {ECO:0000255; 3-HAOxase; 3-hydroxyanthranilate 3,4-dioxygenase; 3-hydroxyanthranilate 3,4-dioxygenase {ECO:0000255; 3-hydroxyanthranilate oxygenase; 3-hydroxyanthranilate oxygenase {ECO:0000255; 3-hydroxyanthranilic acid dioxygenase; 3-hydroxyanthranilic acid dioxygenase {ECO:0000255; Haao; HAD; HAD {ECO:0000255; HAMAP-Rule:MF_03019}; HAO | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 56823 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P46953 | |
| HAAO | |
| Recombinant Rat HAAO/3-HAO Protein (Met1-Gly286). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction