missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAAO Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48755-25ul
This item is not returnable.
View return policy
Description
HAAO Polyclonal antibody specifically detects HAAO in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HAAO | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| 3-HAO, 3-hydroxyanthranilate 3,4-dioxygenase, 3-hydroxyanthranilic acid dioxygenase, EC 1.13.11.6,3-hydroxyanthranilate oxygenase, HAD, HAO | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PLSLFGDTYETQVIAYGQGSSEGLRQNVDVWLWQLEGSSVVTMGGRRLSLAPDDSLLVLAGTSYAWERTQGSVALSVTQDP | |
| 25 μL | |
| Lipid and Metabolism | |
| 23498 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur