missing translation for 'onlineSavingsMsg'
Learn More

HAAO Antibody (4D1), Novus Biologicals™

Product Code. 18388569 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388569 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388569 Supplier Novus Biologicals Supplier No. H00023498M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HAAO Monoclonal antibody specifically detects HAAO in Human, Bacteria samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAAO
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4D1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH29510
Gene Alias 3-HAO, 3-hydroxyanthranilate 3,4-dioxygenase, 3-hydroxyanthranilic acid dioxygenase, EC 1.13.11.6,3-hydroxyanthranilate oxygenase, HAD, HAO
Host Species Mouse
Immunogen HAAO (AAH29510.1, 97 a.a. ~ 196 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ANTVGLVVERRRLETELDGLRYYVGDTMDVLFEKWFYCKDLGTQLAPIIQEFFSSEQYRTGKPIPDQLLKEPPFPLSTRSIMEPMSLDAWLDSHHRELQA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23498
Target Species Human, Bacteria
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.