missing translation for 'onlineSavingsMsg'
Learn More

HA130, MedChemExpress

Product Code. 16463707
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463707

missing translation for 'mfr': Medchem Express HY19329

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HA130 is a selective autotaxin (ATX) inhibitor with an IC50 of 28 nM.

HA130 is a selective autotaxin (ATX) inhibitor with an IC50 of 28 nM.

Specifications

Chemical Name or Material HA130
CAS 1229652-21-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H19BFNO5S
Quantity 50 mg
Solubility Information DMSO : ≥ 39 mg/mL (84.18 mM)
SMILES O=C1N(CC2=CC=C(F)C=C2)C(S/C1=C\C3=CC=C(OCC4=CC(B(O)O)=CC=C4)C=C3)=O
Molecular Weight (g/mol) 463.29
Formula Weight 463.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.