missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HA Tag Polyclonal Antibody, HRP

Description
PA1-985-HRP detects HA tagged proteins in Western Blot applications. This antibody is specific for the HA tag and does not detect other proteins.
HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
Specifications
Specifications
| Antigen | HA Tag |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with proprietary stabilizer and 0.07% Kathon |
| Gene Alias | HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to the residues Y P Y D V P D Y A of the HA epitope of influenza hemagglutinin. |
| Purification Method | Affinity chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?