missing translation for 'onlineSavingsMsg'
Learn More

HA Tag Antibody (16.43), PE/Atto594, Novus Biologicals™

Product Code. 18165738 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18165738 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18165738 Supplier Novus Biologicals Supplier No. NBP250416PEATT594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

HA Tag Monoclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA Tag
Applications Flow Cytometry
Classification Monoclonal
Clone 16.43
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias HA epitope tag, Hemagglutinin
Host Species Rat
Immunogen This HA Tag Antibody (16.43) was developed against the C-terminal influenza virus hemagglutinin (HA) tag, YPYDVPDYA.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Epitope Tags, HA Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.