missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences HA-1H peptide VLHDDLLEA (HLA-A*0201)
Shop All IBA Lifesciences Products

missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
missing translation for 'unitSize'
1mg
Description
Specifications
Specifications
| Content And Storage | Recommended: -25°C to -15°C |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction