missing translation for 'onlineSavingsMsg'
Learn More

HA-100, MedChemExpress

Product Code. 16454496
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454496

Brand: Medchem Express HY100984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HA-100 is a potent protein kinase inhibitor, with IC50s of 4 μM, 8 μM, 12 μM and 240 μM for cGMP-dependent protein kinase (PKG), cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and MLC-kinase, respectively. HA-100 also used as a ROCK inhibitor.

HA-100 is a potent protein kinase inhibitor, with IC50s of 4 μM, 8 μM, 12 μM and 240 μM for cGMP-dependent protein kinase (PKG), cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and MLC-kinase, respectively. HA-100 also used as a ROCK inhibitor.

Specifications

Chemical Name or Material HA-100
CAS 84468-24-6
Color White
Purity Grade Notes Research
Molecular Formula C13H15N3O2S
Quantity 100 mg
Solubility Information DMSO : 150 mg/mL (540.85 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=CC2=C1C=CN=C2)(N3CCNCC3)=O
Molecular Weight (g/mol) 277.34
Formula Weight 277.34
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.