missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA-1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03292-100UL
This item is not returnable.
View return policy
Description
HA-1 Polyclonal specifically detects HA-1 in Human, Rat samples. It is validated for Western Blot.
Specifications
| HA-1 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| histocompatibility (minor) HA-1, HLA-HA1, KIAA0223HA-1, minor histocompatibility antigen HA-1, minor histocompatibility protein HA-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human HA-1 (NP_036424.2). | |
| 100 μL | |
| Signal Transduction | |
| 23526 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction