missing translation for 'onlineSavingsMsg'
Learn More

HA-1 Mouse anti-Nonspecies specific, Clone: 12CA5, Novus Biologicals™

Product Code. 18331162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331162 25 μg 25µg
18391842 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18331162 Supplier Novus Biologicals Supplier No. NBP31188825UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HA-1 Monoclonal antibody specifically detects HA-1 in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 12CA5
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Formulation PBS
Gene Alias histocompatibility (minor) HA-1, HLA-HA1, KIAA0223HA-1, minor histocompatibility antigen HA-1, minor histocompatibility protein HA-1
Host Species Mouse
Immunogen Balb/C mice were immunized with a 9 amino acid petide to influenza virus hemagglutinin (Uniprot: Q3TBD2)
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23526
Target Species Non-species specific
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.