missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HA-1 Antibody (4H7), Novus Biologicals™

Product Code. 18344189 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344189 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344189 Supplier Novus Biologicals™ Supplier No. H00023526M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HA-1 Monoclonal antibody specifically detects HA-1 in Human,Mouse samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen HA-1
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 4H7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH65223.1
Gene Alias histocompatibility (minor) HA-1, HLA-HA1, KIAA0223HA-1, minor histocompatibility antigen HA-1, minor histocompatibility protein HA-1
Host Species Mouse
Immunogen HMHA1 (AAH65223.1, 91 a.a. ∽ 190 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HRSPLTAASPGELPTEGAGPDVVEDISHLLADVARFAEGLEKLKECVLHDDLLEARRPRAHECLGEALRVMHQIISKYPLLNTVETLTAAGTLIAKVKAF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23526
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.