missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ H6PD Recombinant Rabbit Monoclonal Antibody (2I3P1)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558086

 View more versions of this product

Product Code. 30284107

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SDFQVLRAKY RESPLVSAWS EELISKLAND IEATAVRAVR RFGQFHLALS GGSSPVALFQ QLATAHYGFP WAHTHLWLVD ERCVPLSDPE SNFQGLQAHL LQHVRIPYYN IHPMPVHLQQ RLCAEEDQGA QIYAREISAL VANSSFDLVL LGMGADGHTA SLFPQSPTGL DGEQLVVLTT SPSQPHRRMS LSLPLINRAK KVAVLVMGRM KREITTLVSR VGHEPKKWPI SGVLPHSGQL VWYMDYDAFL G.

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

H6PD
Recombinant Monoclonal
1.08 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q8CFX1
H6PD
Recombinant fusion protein containing a sequence corresponding to amino acids 541-791 of human H6PD/GDH (NP_004276.2).
100 μL
Primary
Mouse
Antibody
IgG
ELISA, Immunocytochemistry
2I3P1
Unconjugated
H6PD
6PGL; 6-phosphogluconolactonase; AI785303; CORTRD1; EGK_00198; G6PD, H form; G6pd1; G6PDH; GDH; GDH/6PGL endoplasmic bifunctional protein; glucose 1- dehydrogenase; glucose 1-dehydrogenase; glucose dehydrogenase; glucose dehyrogenase; Glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase 1; glucose-6-phosphate dehydrogenase, salivary; Gpd1; Gpd-1; H6pd; H6pdh; hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase); hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Rabbit
Protein A
RUO
100198
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.