Learn More
Invitrogen™ H6PD Recombinant Rabbit Monoclonal Antibody (2I3P1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558085
Description
Immunogen sequence: SDFQVLRAKY RESPLVSAWS EELISKLAND IEATAVRAVR RFGQFHLALS GGSSPVALFQ QLATAHYGFP WAHTHLWLVD ERCVPLSDPE SNFQGLQAHL LQHVRIPYYN IHPMPVHLQQ RLCAEEDQGA QIYAREISAL VANSSFDLVL LGMGADGHTA SLFPQSPTGL DGEQLVVLTT SPSQPHRRMS LSLPLINRAK KVAVLVMGRM KREITTLVSR VGHEPKKWPI SGVLPHSGQL VWYMDYDAFL G.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.
Specifications
| H6PD | |
| Recombinant Monoclonal | |
| 1.08 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q8CFX1 | |
| H6PD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 541-791 of human H6PD/GDH (NP_004276.2). | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 2I3P1 | |
| Unconjugated | |
| H6PD | |
| 6PGL; 6-phosphogluconolactonase; AI785303; CORTRD1; EGK_00198; G6PD, H form; G6pd1; G6PDH; GDH; GDH/6PGL endoplasmic bifunctional protein; glucose 1- dehydrogenase; glucose 1-dehydrogenase; glucose dehydrogenase; glucose dehyrogenase; Glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase 1; glucose-6-phosphate dehydrogenase, salivary; Gpd1; Gpd-1; H6pd; H6pdh; hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase); hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100198 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.