missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H4K5me1 Recombinant Rabbit Monoclonal Antibody (RM457)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA562168
This item is not returnable.
View return policy
Description
This antibody reacts to Histone H4 monomethylated at Lysine 5 (K5me1). No cross reactivity with non-modified Lysine 5, or other monomethylations in histone H4. Predicted to react with all vertebrates.
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
| H4K5me1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| P62805 | |
| HIST1H4A | |
| A monomethyl-peptide corresponding to monomethyl-Histone H4 (Lys5). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Luminex | |
| RM457 | |
| Unconjugated | |
| HIST1H4A | |
| H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4-16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; HIST2H4; HIST2H4A; HIST2H4B; HIST4H4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 121504 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction