missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H4/n Antibody (7A2E10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | H4/n |
|---|---|
| Clone | 7A2E10 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18289270
|
Novus Biologicals
NBP2-52489 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680367
|
Novus Biologicals
NBP2-52489-0.025mg |
0.025 mg |
£161.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
H4/n Monoclonal specifically detects H4/n in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.Specifications
| H4/n | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| 8370 | |
| Synthesized peptide of human HIST2H4A (AA: GGAKRHRK(Me3)VLRDNIQ) . | |
| Primary | |
| 11.4 kDa |
| 7A2E10 | |
| Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| HIST2H4A | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title