missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H4/n [ac Lys16] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £549.00
Specifications
| Antigen | H4/n |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233111
|
Novus Biologicals
NBP3-35924-100ul |
100 μL |
£549.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228154
|
Novus Biologicals
NBP3-35924-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
H4/n Polyclonal antibody specifically detects H4/n in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| H4/n | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FO108, H4, H4 histone family, member N, H4 histone, family 2, H4/A, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4F2H4/B, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FNH4/C, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4B, HIST4H4, histone 2, H4, histone 2, H4a, histone cluster 2, H4a, histone H4, histone IV, family 2 | |
| A synthetic acetylated peptide around K16 of human H4/n (NP_003539.1).,, Sequence:, GAKRH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 8370 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title