missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H4 Clustered Histone 1 [ac Lys5] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £549.00
Specifications
| Antigen | H4 Clustered Histone 1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 5ug antibody for 5ug-10ug of Chromatin, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ChIP Assay, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231494
|
Novus Biologicals
NBP3-35514-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232358
|
Novus Biologicals
NBP3-35514-100ul |
100 μL |
£549.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
H4 Clustered Histone 1 Polyclonal antibody specifically detects H4 Clustered Histone 1 in Human,Mouse,Rat samples. It is validated for ChIP assay,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| H4 Clustered Histone 1 | |
| ChIP Assay, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| H4-16;H4C11;H4C12;H4C13;H4C14;H4C15;H4C2;H4C3;H4C4;H4C5;H4C6;H4C8;H4C9;H4FA, H4F4, HIST1H4A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human H4 Clustered Histone 1 (NP_001029249.1).,, Sequence:, GRGKG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 5ug antibody for 5ug-10ug of Chromatin, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 8359 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title