Learn More
H3K27cr Recombinant Polyclonal Antibody (9HCLC), ChIP-Verified, Invitrogen™
Rabbit Recombinant Polyclonal Antibody
£110.00 - £508.00
Specifications
| Antigen | H3K27cr |
|---|---|
| Clone | 9HCLC |
| Concentration | 0.5 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This antibody is predicted to react with Rat, Bovine. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.Specifications
| H3K27cr | |
| 0.5 mg/mL | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P68431, P68433 | |
| H3C1 | |
| IgG | |
| Protein A | |
| Antibody |
| 9HCLC | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| 360198, 8350 | |
| Peptide corresponding to human Crotonyl-Histone H3.1 (Lys27) [ aa23-aa33] | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| H3C1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.