missing translation for 'onlineSavingsMsg'
Learn More

H2L5186303, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001621
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001621

Brand: Tocris Bioscience 4878/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective LPA2 receptor antagonist

Potent and selective lysophosphatidic acid 2 (LPA2) receptor antagonist (IC50 values are 8.9, 1230 and 27354 nM for LPA2, LPA3 and LPA1 receptors respectively, in a LPA-elicited calcium mobilization assay).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139262-76-3
Molecular Formula C26H20N2O8
Molecular Weight (g/mol) 488.452
InChI Key HZFPOTBCYPWQSH-DZDAAMPGSA-N
Synonym z,z-4,4'-1,3-phenylenebis oxy-4,1-phenyleneimino bis 4-oxo-2-butenoic acid, 2z-3-4-3-4-2z-3-carboxyprop-2-enamido phenoxy phenoxy phenyl carbamoyl prop-2-enoic acid
PubChem CID 90488981
IUPAC Name (Z)-4-[4-[3-[4-[[(Z)-3-carboxyprop-2-enoyl]amino]phenoxy]phenoxy]anilino]-4-oxobut-2-enoic acid
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)NC(=O)C=CC(=O)O)OC3=CC=C(C=C3)NC(=O)C=CC(=O)O

Specifications

Quantity 10 mg
Formula Weight 488.45
Percent Purity >98%
Chemical Name or Material H2L5186303
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.