missing translation for 'onlineSavingsMsg'
Learn More

H2L 5765834, Tocris Bioscience™

Product Code. 30001686
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001686

Brand: Tocris Bioscience 4870/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LPA1,3,5 antagonist

Antagonist of the lysophosphatidic acid receptors LPA1, LPA5 and LPA3 (IC50 values are 94, 463 and 752 nM respectively). Exhibits no effect at LPA2 or LPA4 receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420841-84-5
Molecular Formula C21H12N2O7
Molecular Weight (g/mol) 404.334
InChI Key HFYPTENHTPNXGP-UHFFFAOYSA-N
Synonym 2-3-4-nitrophenoxy phenyl-1,3-dioxoisoindole-5-carboxylic acid, 2-3-4-nitrophenoxy phenyl-1,3-dioxoisoindoline-5-carboxylic acid, 2,3-dihydro-2-3-4-nitrophenoxy phenyl-1,3-dioxo-1h-isoindole-5-carboxylic acid, 2-3-4-nitrophenoxy phenyl-1,3-dioxo-2,3-dihydro-1h-isoindole-5-carboxylic acid
PubChem CID 1365686
IUPAC Name 2-[3-(4-nitrophenoxy)phenyl]-1,3-dioxoisoindole-5-carboxylic acid
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)[N+](=O)[O-])N3C(=O)C4=C(C3=O)C=C(C=C4)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 404.33
Percent Purity >99%
Chemical Name or Material H2L 5765834
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.