missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H2BFWT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563280
This item is not returnable.
View return policy
Description
Immunogen sequence: PKEANSTTSQ KQSKQRKRGR HGPRRCHSNC RGDSFATYFR RVL Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 39%.
Atypical histone H2B. Nucleosomes containing it are structurally and dynamically indistinguishable from those containing conventional H2B. However, unlike conventional H2B, does not recruit chromosome condensation factors and does not participate in the assembly of mitotic chromosomes. May be important for telomere function.
Specifications
| H2BFWT | |
| Polyclonal | |
| Unconjugated | |
| H2BW1 | |
| H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member W, testis-specific; H2BFWT; Histone H2B type W-T | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z2G1 | |
| H2BW1 | |
| Recombinant protein corresponding to Human H2BFWT. Recombinant protein control fragment (Product #RP-100384). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction