missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2AFV Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £366.00
Specifications
| Antigen | H2AFV |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18393014
|
Novus Biologicals
NBP3-04426-20UL |
20 μL |
£153.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18386016
|
Novus Biologicals
NBP3-04426-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
H2AFV Polyclonal specifically detects H2AFV in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| H2AFV | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 94239 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ26479, H2A histone family, member V, H2A.F/Z, H2AV, histone H2A.F/Z, histone H2A.V, MGC10170, MGC10831, MGC1947, purine-rich binding element protein B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human H2AFV (NP_036544.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title