missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H2A.ZK7ac Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112438
This item is not returnable.
View return policy
Description
H2A.ZK7ac Polyclonal Antibody for Western Blot, ICC/IF, ELISA, IP, ChIP
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.
Specifications
| H2A.ZK7ac | |
| Polyclonal | |
| Unconjugated | |
| H2AZ1 | |
| 11ac; Acetyl-Histone H2A.Z K7; Acetyl-Histone H2A.Z Lys7; H2A histone family member Z; H2A histone family, member Z; H2A.z; H2A.Z-1; H2A.ZK5; H2A.ZK5ac; H2A.ZLys7ac; H2A/z; H2AFZ; H2AFZac; H2AZ; H2AZ histone; H2AZK11ac; H2AZK7ac; H2AZLys7ac; histone H2A.Z | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3015, 94239 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0C0S5, Q71UI9 | |
| H2AZ1, H2az2 | |
| Peptide sequence around site of Acetyl-Lys (7) derived from Human Histone H2A.Z (2-14aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction