missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: ALPVTTAEGM AKKVTKAGGS AALSPSKKRK NSKKKNQPGK YSQLVVETIR RLGERNGSSL AKIYTEAKKV PWFDQQNG Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.
Histones H1 are necessary for the condensation of nucleosome chains into higher-order structures.
Specifications
Specifications
| Antigen | H1FX |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | H1-10 |
| Gene Accession No. | Q92522 |
| Gene Alias | H1 histone family member X; H1 histone family, member X; H1.10; H1FX; H1X; Histone H1.X; histone H1x |
| Gene Symbols | H1-10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
