missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ H1FX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29518
This item is not returnable.
View return policy
Description
H1FX Polyclonal antibody specifically detects H1FX in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| H1FX | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8971 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| H1 histone family, member X, H1X, histone H1x, MGC15959, MGC8350 | |
| Between 163 and 213 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction