missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ H1FOO 293T Cell Transient Overexpression Lysate

Product Code. 18485609 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18485609 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18485609 Supplier Novus Biologicals™ Supplier No. H00132243T02
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Research Category Phospho Specific
Target H1FOO
Synonym H1 histone family, member O, oocyte-specific, H1OO, histone H1oo, MGC50807, Oocyte-specific histone H1, Oocyte-specific linker histone H1, osH1
Gene Symbol H1FOO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.