missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H1F0 Antibody (HH1/1784R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54459CL1
This item is not returnable.
View return policy
Description
H1F0 Monoclonal antibody specifically detects H1F0 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| H1F0 | |
| Monoclonal | |
| CoraFluor 1 | |
| H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0 | |
| Recombinant full-length human H1F0 protein | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Epigenetics, Nuclear Envelope Markers | |
| 3005 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), CyTOF | |
| HH1/1784R | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction