missing translation for 'onlineSavingsMsg'
Learn More

H-VAL-TYR-OH, TRC

Product Code. 30455910
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455910 10 mg 10mg
30455441 100 mg -
30451240 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455910 Supplier Toronto Research Chemicals Supplier No. TRCV99173810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3061-91-4
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
Synonym L-Tyrosine, N-L-valyl-, Tyrosine, N-L-valyl-, L- (7CI,8CI), Tyrosine, N-valyl- (6CI), L-Valyl-L-tyrosine, N-L-Valyl-L-tyrosine, Valyltyrosine
IUPAC Name (2S)-2-[[(2S)-2-amino-3-methylbutanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(C)[C@H](N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O

Specifications

InChI Formula InChI=1S/C14H20N2O4/c1-8(2)12(15)13(18)16-11(14(19)20)7-9-3-5-10(17)6-4-9/h3-6,8,11-12,17H,7,15H2,1-2H3,(H,16,18)(H,19,20)/t11-,12-/m0/s1
Quantity 10 mg
Formula Weight 280.1423
Shipping Condition Room Temperature
Chemical Name or Material H-VAL-TYR-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.