missing translation for 'onlineSavingsMsg'
Learn More

H-VAL-TYR-OH, TRC

Product Code. 30451240
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451240 50 mg -
30455441 100 mg -
30455910 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451240 Supplier Toronto Research Chemicals Supplier No. TRCV99173850MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3061-91-4
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
Synonym L-Tyrosine, N-L-valyl-, Tyrosine, N-L-valyl-, L- (7CI,8CI), Tyrosine, N-valyl- (6CI), L-Valyl-L-tyrosine, N-L-Valyl-L-tyrosine, Valyltyrosine
IUPAC Name (2S)-2-[[(2S)-2-amino-3-methylbutanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(C)[C@H](N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O

Specifications

InChI Formula InChI=1S/C14H20N2O4/c1-8(2)12(15)13(18)16-11(14(19)20)7-9-3-5-10(17)6-4-9/h3-6,8,11-12,17H,7,15H2,1-2H3,(H,16,18)(H,19,20)/t11-,12-/m0/s1
Quantity 50 mg
Formula Weight 280.1423
Shipping Condition Room Temperature
Chemical Name or Material H-VAL-TYR-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.