missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-Val-Gln-OH, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | H-Val-Gln-OH |
| CAS | 42854-54-6 |
| Molecular Formula | C10 H19 N3 O4 |
| Quantity | 10 mg |
| Synonym | L-Glutamine','L-valyl- (9CI','ACI), L-Glutamine','N2-L-valyl- (ZCI), L-Valyl-L-glutamine (ACI), Glutamine','N2-L-valyl- (7CI), 113: PN: WO2011146121 PAGE: 116 claimed sequence, 27: PN: EP2161028 PAGE: 10 claimed protein, 471: PN: WO2005081628 SEQID: 1580 claimed protein, 8: PN: JP2007217358 SEQID: 8 claimed protein, 98: PN: US20070066537 PAGE: 17 claimed protein |
| InChI Formula | InChI=1S/C10H19N3O4/c1-5(2)8(12)9(15)13-6(10(16)17)3-4-7(11)14/h5-6,8H,3-4,12H2,1-2H3,(H2,11,14)(H,13,15)(H,16,17)/t6-,8-/m0/s1 |
| SMILES | CC(C)[C@H](N)C(=O)N[C@@H](CCC(=O)N)C(=O)O |
| IUPAC Name | (2S)-5-amino-2-[[(2S)-2-amino-3-methyl-butanoyl]amino]-5-oxo-pentanoic acid |
| Molecular Weight (g/mol) | 245.276 |
| Formula Weight | 245.1376 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction