missing translation for 'onlineSavingsMsg'
Learn More

H-Tyr(3-I)-OH, MedChemExpress

Product Code. 16435447
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16435447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16435447 Supplier Medchem Express Supplier No. HYW008452
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress H-Tyr(3-I)-OH is a potent and effective tyrosine hydroxylase inhibitor. H-Tyr(3-I)-OH is an intermediate in the production of thyroid hormones and has a role as a human or mouse metabolite.

H-Tyr(3-I)-OH is a potent and effective tyrosine hydroxylase inhibitor. H-Tyr(3-I)-OH is an intermediate in the production of thyroid hormones and has a role as a human or mouse metabolite.

Specifications

Chemical Name or Material H-Tyr(3-I)-OH
CAS 70-78-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H10INO3
Quantity 50 mg
Solubility Information H2O : 5 mg/mL (16.28 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (3.26 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](N)CC1=CC=C(O)C(I)=C1
Molecular Weight (g/mol) 307.09
Formula Weight 307.09
Percent Purity 98.86%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.