missing translation for 'onlineSavingsMsg'
Learn More

H-Trp-Phe-OH, TRC

Product Code. 30403020
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403020 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403020 Supplier Toronto Research Chemicals Supplier No. TRCH2186051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material H-Trp-Phe-OH
CAS 6686-02-8
Molecular Formula C20 H21 N3 O3
Quantity 1 g
Synonym L-Tryptophyl-L-phenylalanine, 14: PN: JP2014079213 TABLE: 2 claimed sequence, 2: PN: WO2013070776 PAGE: 109 claimed sequence, Tryptophanylphenylalanine, Tryptophylphenylalanine
InChI Formula InChI=1S/C20H21N3O3/c21-16(11-14-12-22-17-9-5-4-8-15(14)17)19(24)23-18(20(25)26)10-13-6-2-1-3-7-13/h1-9,12,16,18,22H,10-11,21H2,(H,23,24)(H,25,26)/t16-,18-/m0/s1
SMILES N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](Cc3ccccc3)C(=O)O
IUPAC Name (2S)-2-[[(2S)-2-amino-3-(1H-indol-3-yl)propanoyl]amino]-3-phenylpropanoic acid
Molecular Weight (g/mol) 351.399
Formula Weight 351.158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.