Learn More
Invitrogen™ H-Ras Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588424
Description
Immunogen sequence: MTEYKLVVVG AGGVGKSALT IQLIQNHFVD EYDPTIEDSY RKQVVIDGET CLLDILDTAG QEEYSAMRDQ YMRTGEGFLC VFAINNTKSF EDIHQYREQI KRVKDSDDVP MVLVGNKCDL AARTVESRQA QDLARSYGIP YIETSAKTRQ GVEDAFYTLV REIRQHKLRK LNPPDESGPG CMSCKCVLS; Positive Samples: Mouse brain; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Golgi apparatus, Golgi apparatus membrane, Lipid-anchor, Lipid-anchor, Nucleus, perinuclear region.
Monomeric G protein Ras functions as a molecular switch linking receptor and nonreceptor tyrosine kinase activation to a number of downstream signaling pathways leading to distinct cytoplasmic or nuclear events. Each mammalian cell contains three Ras proto-oncogene coding for closely related Ras proteins: H-, K-, N-Ras. Oncogenic mutations in Ras genes are present in ~30% of all human cancers. Constitutive activation of Ras due to mutations or overexpression stimulates proliferation and inhibition of apoptosis. K-Ras mutations are common in pancreatic, colorectal and nonsmall-cell lung carcinomas; H-Ras mutations are common in bladder, kidney and thyroid carcinomas; N-Ras mutations are found in melanoma, hematological malignancies and hepatocellular carcinaoma.
Specifications
| H-Ras | |
| Polyclonal | |
| Unconjugated | |
| HRAS | |
| C-BAS/HAS; c-Ha-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; C-HA-RAS1; c-has/bas p21 protein; c-H-ras; c-rasHa; c-ras-Ki-2 activated oncogene; CTLO; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas, N-terminally processed; GTPase Hras1; HAMSV; Ha-ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene; Harvey rat sarcoma virus oncogene 1; Harvey-ras; Hras; H-ras; H-ras 1 protein; HRas proto-oncogene, GTPase; HRAS1; H-Ras-1; Hras-1; H-RASIDX; Kras2; p19 H-RasIDX protein; p21ras; ras; Ras family small GTP binding protein H-Ras; ras p21; RASH1; transformation gene: oncogene HAMSV; transforming protein P21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15461, 3265 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.03 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P01112, Q61411 | |
| HRAS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human HRAS (NP_0053341). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.