missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress H-Phe-Phe-OH

Product Code. 16459326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459326 Supplier MedChemExpress Supplier No. HYW007970

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress H-Phe-Phe-OH is a peptide made of two phenylalanine molecules; Phenylalanine is an essential amino acid and the precursor for the amino acid tyrosine.

Specifications

Purity 97.0%
CAS 2577-40-4
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Neuroscience-Neurodegeneration
Molecular Weight (g/mol) 312.36 g/mol
Product Type Natural Products
Quantity 100 mg
Shipping Condition Room temperature
Solubility 80% Acetic acid/water : 50 mg/mL (160.07 mM∣Need ultrasonic) ∣DMSO : <1 mg/mL (ultrasonic∣warming∣heat to 60°C) (insoluble or slightly soluble) ∣H2O : < 0.1 mg/mL (ultrasonic∣warming∣heat to 60°C) (insoluble)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.