missing translation for 'onlineSavingsMsg'
Learn More

H-Leu-Leu-Tyr-OH, TRC

Product Code. 30391600
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391600 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391600 Supplier Toronto Research Chemicals Supplier No. TRCH24690525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material H-Leu-Leu-Tyr-OH
CAS 20368-24-5
Molecular Formula C21 H33 N3 O5
Quantity 25 mg
Synonym L-Leucyl-L-leucyl-L-tyrosine, 10: PN: WO2013116663 SEQID: 6 Unclaimed Protein, 2: PN: WO2009046832 PAGE: 100 Claimed Protein, Leucylleucyltyrosine
InChI Formula InChI=1S/C21H33N3O5/c1-12(2)9-16(22)19(26)23-17(10-13(3)4)20(27)24-18(21(28)29)11-14-5-7-15(25)8-6-14/h5-8,12-13,16-18,25H,9-11,22H2,1-4H3,(H,23,26)(H,24,27)(H,28,29)/t16-,17-,18-/m0/s1
SMILES CC(C)C[C@H](N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O
IUPAC Name (2S)-2-[[(2S)-2-[[(2S)-2-amino-4-methylpentanoyl]amino]-4-methylpentanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 407.504
Formula Weight 407.242
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.