missing translation for 'onlineSavingsMsg'
Learn More

H-D-Trp-OH, MedChemExpress

Product Code. 16493429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493429 Supplier Medchem Express Supplier No. HYW012479
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress H-D-Trp-OH is a D-stereoisomer of tryptophan and occasionally found in naturally produced peptides such as the marine venom peptide.

H-D-Trp-OH is a D-stereoisomer of tryptophan and occasionally found in naturally produced peptides such as the marine venom peptide.

Specifications

Chemical Name or Material H-D-Trp-OH
CAS 153-94-6
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C11H12N2O2
Solubility Information DMSO : 3.33 mg/mL (16.31 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)[C@H](N)CC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 204.23
Quantity 1 g
Formula Weight 204.23
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.