missing translation for 'onlineSavingsMsg'
Learn More

H-Arg-4MβNA, MedChemExpress

Product Code. 16470967
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470967 50 mg 50mg
16464678 10 mg 10mg
16423196 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16470967 Supplier Medchem Express Supplier No. HY112688
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis.

H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis.

Specifications

Chemical Name or Material H-Arg-4MβNA
CAS 60285-94-1
Color White
Purity Grade Notes Research
Molecular Formula C17H23N5O2
Quantity 50 mg
Solubility Information H2O : ≥ 125 mg/mL (379.48 mM)
SMILES COC1=CC(NC([C@@H](N)CCCNC(N)=N)=O)=CC2=CC=CC=C12
Molecular Weight (g/mol) 329.4
Formula Weight 329.4
Percent Purity 99.86%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.