missing translation for 'onlineSavingsMsg'
Learn More

H-7 dihydrochloride, Thermo Scientific Chemicals

Product Code. 15474239
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474239 10 mg 10mg
15484239 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474239 Supplier Thermo Scientific Alfa Aesar Supplier No. J61697.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein kinase inhibitor. Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Protein kinase inhibitor
  • Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108930-17-2
Molecular Formula C14H19Cl2N3O2S
Molecular Weight (g/mol) 364.285
MDL Number MFCD00036961
InChI Key OARGPFMFRLLKPF-UHFFFAOYSA-N
Synonym h-7 dihydrochloride, h-7, dihydrochloride, 1-5-isoquinolinylsulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine, 2hcl, 1-5-isoquinoline-sulfonyl-2-methylpiperazine, 5-2-methylpiperazin-1-ylsulfonyl isoquinoline dihydrochloride, isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride, 1-5-isoquinolylsulfonyl-2-methylpiperazine, chloride, chloride, 1-5-isoquinolinesulfonyl-2-methyl-piperazine dihydrochloride
PubChem CID 73332
ChEBI CHEBI:82739
IUPAC Name 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride
SMILES CC1CNCCN1S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl

Specifications

Melting Point 191°C to 193°C
Color White
Quantity 10 mg
Beilstein 5840763
Formula Weight 364.29
Physical Form Powder
Chemical Name or Material H-7 dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.