missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ H-7 dihydrochloride
GREENER_CHOICE

Product Code. 18084052
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18084052 50 mg 50mg
18094802 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084052 Supplier Tocris Bioscience™ Supplier No. 0542/50
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Protein kinase inhibitor

H-7 dihydrochloride is a protein kinase inhibitor (IC50 values for inhibition of PKA, PKG, PKC and myosin light chain kinase are 3.0, 5.8, 6.0 and 97.0 μM, respectively. H-7 shows antiviral activity against influenza A (IC50 = 10 μM) in vitro. H-7 also inhibits interleukin-stimulated secretion of IgM and blocks PMA -stimulated interleukin 1β production.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material '±-1-(5-Isoquinolinesulphonyl)-2-methylpiperazine dihydrochloride
CAS 108930-17-2
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C14H17N3O2S.2HCl
Formula Weight Observed MW: 364.29

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.