missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H-2Kb/H-2Db Monoclonal Antibody (5041.16.1), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517999
This item is not returnable.
View return policy
Description
MA5-17999 targets H-2KbDb in FACS applications and shows reactivity with Mouse samples. The MA5-17999 immunogen is mouse H-2KbDb.
H-2KbDb monoclonal antibody is specific for cells expressing the H-2K antigen coded for by the b haplotype, and for cells expressing the H-2D antigen coded for by the b haplotype. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| H-2Kb/H-2Db | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P01901, P03991, P04223, P14428 | |
| H2-K1 | |
| Mouse H-2KbDb. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 5041.16.1 | |
| FITC | |
| H2-K1 | |
| class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H2DB; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; H2KB; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen | |
| Mouse | |
| Protein G | |
| RUO | |
| 14972 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction