missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ H 151
GREENER_CHOICE

Product Code. 18717606
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18717606

Brand: Tocris Bioscience™ 6675/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

STING antagonist

H 151 is a STING antagonist. Covalently binds STING at Cys91, blocking palmitoylation and preventing assembly of STING clusters. Decreases inflammatory cytokine production in human and mouse cells in vitro. Also blocks CMA (Cat. No. 6677) -induced increases in levels of inflammatory cytokines in mice, and alleviates systemic inflammation in a mouse model of autoinflammatory disease.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Ethylphenyl)-N'-1H-indol-3-yl-urea
CAS 941987-60-6
Synonym ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
Quantity 10 mg
Target STING Antagonists
Molecular Formula C17H17N3O
Formula Weight 279.34
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.