missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GYS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22371-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene, liver glycogen synthase, catalyzes the rate-limiting step in the synthesis of glycogen - the transfer of a glucose molecule from UDP-glucose to a terminal branch of the glycogen molecule. Mutations in this gene cause glycogen storage disease type 0 (GSD-0) - a rare type of early childhood fasting hypoglycemia with decreased liver glycogen content.Specifications
| GYS2 | |
| Polyclonal | |
| Unconjugated | |
| GYS2 | |
| glycogen synthase 2 (liver), GYS2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 232493, 25623, 2998 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P17625, P54840, Q8VCB3 | |
| Gys2 | |
| GYS2 Fusion Protein Ag18012 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction