missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GYS2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588921

Product Code. 16546991

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DLLDWRYLGR YYQHARHLTL SRAFPDKFHV ELTSPPTTEG FKYPRPSSVP PSPSGSQASS PQSSDVEDEV EDERYDEEEE AERDRLNIKS PFSLSHVPHG KKKLHGEYKN; Positive Samples: Mouse liver.

The protein encoded by this gene, liver glycogen synthase, catalyzes the rate-limiting step in the synthesis of glycogen - the transfer of a glucose molecule from UDP-glucose to a terminal branch of the glycogen molecule. Mutations in this gene cause glycogen storage disease type 0 (GSD-0) - a rare type of early childhood fasting hypoglycemia with decreased liver glycogen content.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GYS2
Polyclonal
Unconjugated
GYS2
BC021322; cb765; Glycogen [starch] synthase, liver; glycogen liver; glycogen synthase; glycogen synthase 2; glycogen synthase 2 (liver); glycogen synthase, liver; GLYSN; GYS2; LGS; zgc:112057
Rabbit
Affinity Chromatography
RUO
232493
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.6 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q8VCB3
GYS2
Recombinant fusion protein containing a sequence corresponding to amino acids 594-703 of human GYS2 (NP_0687762).
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.