missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GYS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599507
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Glycogen Synthase.
Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| GYS1 | |
| Polyclonal | |
| Unconjugated | |
| Gys1 | |
| EC 2.4.1.11; glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 3, brain; GSY; GYS; GYS1; Gys3; MGS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14936, 2997, 690987 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2RRU1, P13807, Q9Z1E4 | |
| Gys1 | |
| A synthesized peptide derived from human GYS1(Accession P13807), corresponding to amino acid residues E619-E669. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction