missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GYS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583566
This item is not returnable.
View return policy
Description
Immunogen sequence: PSLSRHSSPH QSEDEEDPRN GPLEEDGERY DEDEEAAKDR RNIRAPEWPR RASCTSSTSG SKRNSVDTAT SSSLSTPSE Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 86%.
Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| GYS1 | |
| Polyclonal | |
| Unconjugated | |
| Gys1 | |
| EC 2.4.1.11; glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 3, brain; GSY; GYS; GYS1; Gys3; MGS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2997 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P13807 | |
| Gys1 | |
| Recombinant protein corresponding to Human GYS1. Recombinant protein control fragment (Product #RP-98893). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction